Learn More
OHAUS Clip Plate-Hybridization Tube
Catalog No. 01804371 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Shaker Attachment
Catalog No. Product Type
01-804-371 Shaker Attachment
1 options
Catalog No. 01-804-371 Supplier OHAUS Supplier No. 83041377
Add to Cart
Add to Cart
This can be attached to the Magnetic Platform for use with the Roto-Shake Genie, Rotator Genie, Enviro-Genie or Incubator-Genie.
  • Pack of 2 metal clip plates to accommodate one 35 mm ID Hybridization Tube up to 300 mm in length.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Shaker Attachment