Learn More
New England Biolabs, Inc. CLIP-Surface™ 488 – 50 nmol
SDP
Catalog No. 508112086
Encompass
CLIP-Surface 488 is a photostable green fluorescent substrate that can be used to label CLIP-tag fusion proteins on the surface of living cells or in vitro.
Catalog No. 50-811-2086 Supplier New England Biolabs, Inc. Supplier No. S9232S
CLIP-Surface 488 is a photostable green fluorescent substrate that can be used to label CLIP-tag fusion proteins on the surface of living cells or in vitro.