Learn More
New England Biolabs, Inc. CLIP-Surface™ 647 – 50 nmol
SDP
Catalog No. 508112088
Encompass
CLIP-Surface 647 is a photostable red fluorescent substrate that can be used to label CLIP-tag fusion proteins on the surface of living cells, or in vitro.
Catalog No. 50-811-2088 Supplier New England Biolabs, Inc. Supplier No. S9234S
CLIP-Surface 647 is a photostable red fluorescent substrate that can be used to label CLIP-tag fusion proteins on the surface of living cells, or in vitro.