Learn More
Aviva Systems Biology Corp CLIP1 Antibody - C-terminal region (ARP61766_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61766_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CLIP1. It was validated on Western Blot. . Alternate names: CLIP, CLIP-170, CLIP170, CYLN1, MGC131604, RSNApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-287-462