Learn More
Aviva Systems Biology Corp Clip3 Antibody - N-terminal region (ARP55276_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55276_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Clip3. It was validated on Western Blot. . Alternate names: 1500005P14Rik, AI844915, Clipr-59ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-021