Learn More
Leica Biosystems CLIPPED GREEN SLIDE 1440/CS
Catalog No. NC1718200
Encompass_Preferred
Snowcoat Clipped Corner Slides are clipped at 45° angles to help reduce glass breakage and fragmentation when loaded into slide printers. These slides have been optimized for use with the Leica printer.
This item is not returnable. View return policy
Catalog No. NC1718200 Supplier Leica Biosystems Supplier No. 3800383
Snowcoat Clipped Corner Slides are clipped at 45° angles to help reduce glass breakage and fragmentation when loaded into slide printers. These slides have been optimized for use with the Leica printer.