Learn More
Med Vet International No. 40 Clipper Blade, Ceramic Edges
Catalog No. 502829975
Encompass_Preferred
Ceramic Edge™ blades feature a certified zirconium oxide ceramic cutter blade that offers the following advantages: Higher durability, stays sharper longer Cutter runs 75% cooler than steel Feature a smoother, faster cutting feel regardless of length or density Break resistant material Fit on Andis models AG, AGC, AGR+ and MBG, Oster A5 and most detachable-type clippers Size: 40
Catalog No. 50-282-9975 Supplier Med Vet International Supplier No. AN40
Ceramic Edge™ blades feature a certified zirconium oxide ceramic cutter blade that offers the following advantages: Higher durability, stays sharper longer Cutter runs 75% cooler than steel Feature a smoother, faster cutting feel regardless of length or density Break resistant material Fit on Andis models AG, AGC, AGR+ and MBG, Oster A5 and most detachable-type clippers Size: 40