Learn More
Med Vet International 3M Rechargeable Surgical Clippers 120 Minutes, 9681, 4/Case
Catalog No. 502832118
Encompass_Preferred
Single blade for standard and trauma procedures Removes all types of hair in a single pass 120 minutes of use time on a minimal 4-hour recharge
Catalog No. 50-283-2118 Supplier Med Vet International Supplier No. 1195679CS
Single blade for standard and trauma procedures Removes all types of hair in a single pass 120 minutes of use time on a minimal 4-hour recharge