Learn More
Grainger Scientific Industries Clip Plates, 15-17 mm Tubes, 6-Piece Set
Catalog No. 502768777
Encompass_Preferred
These clip plates are designed to provide secure, stable support for medium-diameter tubes during laboratory procedures such as shaking, rocking, or incubation. Compatible with common platform systems, the set includes six high-capacity plates that ensure consistent sample positioning even under vigorous agitation, making it an effective solution for high-throughput workflows.
  • Holds up to six 15 mm to 17 mm diameter tubes per plate
  • Ensures secure retention of samples during laboratory agitation
  • Designed for use with common platform systems
  • Includes six plates per set
Catalog No. 50-276-8777 Supplier Grainger Supplier No. 13R345
These clip plates are designed to provide secure, stable support for medium-diameter tubes during laboratory procedures such as shaking, rocking, or incubation. Compatible with common platform systems, the set includes six high-capacity plates that ensure consistent sample positioning even under vigorous agitation, making it an effective solution for high-throughput workflows.
  • Holds up to six 15 mm to 17 mm diameter tubes per plate
  • Ensures secure retention of samples during laboratory agitation
  • Designed for use with common platform systems
  • Includes six plates per set