Description
Utilize Thermo Scientific™ ClipTip™ Pipette Tip Racks, available in three sizes.
- Small Rack: For use with ClipTip 20 and 50 Pipette Tips
- Medium Rack: For use with ClipTip 200 and 300 Pipette Tips
- Large Rack: For use with ClipTip 1000 Pipette Tips
Ordering Information:
Racks are supplied without tips and inserts
Specifications
Specifications
| Description | ClipTip Empty Rack, Small |
| Model | ClipTip Empty Rack |
| For Use With (Equipment) | Thermo Scientific E1-ClipTip |
| Product Line | ClipTip |