Learn More
Carnabio Usa Inc CLK1/10UG/CATALYTIC
Supplier:  Carnabio Usa Inc 0412610
Encompass
CLK1, Catalytic domain, Wild type, Amino Acid 129-484, NP_004062.2, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0737