Learn More
Medchemexpress LLC HY-103082 5mg Medchemexpress, CLK1-IN-1 CAS:2123491-32-5 Purity:>98%
SDP
Catalog No. 501968028
Encompass_Preferred
Medchemexpress, HY-103082 5mg CLK1-IN-1 CAS:2123491-32-5 CLK1-IN-1 is a potent and selective of Cdc2-like kinase 1 (CLK1) inhibitor, with an IC50 of 2 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8028 Supplier Medchemexpress LLC Supplier No. HY1030825MG
Medchemexpress, HY-103082 5mg CLK1-IN-1 CAS:2123491-32-5 CLK1-IN-1 is a potent and selective of Cdc2-like kinase 1 (CLK1) inhibitor, with an IC50 of 2 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.