Learn More
Carnabio Usa Inc CLK3/5UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 041285
Encompass
CLK3, Full-length, Wild type, Amino Acid 1-490, AAH02555.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0384