Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CLK3, GST Tag Recombinant Protein

Catalog No. PV3826
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV3826 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV3826 Supplier Invitrogen™ Supplier No. PV3826
Only null left
Add to Cart
Add to Cart

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.

CLK3 is a dual specificity kinase that can interact with and cause the nuclear redistribution of SR proteins. Two transcript variants encoding different isoforms have been found for CLK3. Related pseudogenes are located on chromosomes 1 and 9.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P49761
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 150mM NaCl, 0.5mM EDTA, 2mM DTT, 0.02% Triton X-100, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 1198
Molecular Weight (g/mol) 86 kDa
Name Human CLK3, GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias AI256811; CDC like kinase 3; cdc2/CDC28-like protein kinase 3; CDC-like kinase 3; Clk3; Dual specificity protein kinase CLK3; PHCLK3; PHCLK3/152; Protein kinase CLK3
Common Name CLK3
Gene Symbol CLK3
Kinase Family CLK Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical phospho-specific activity (no antibody)
Kinase Group Ser/Thr Kinases: CMCG Group
Shipping Condition Dry Ice
System Compatibility Zin-LYTE™, Omnia™ Bulk Substrate
Target Entry CLK3
Target Gene CDC-like kinase 3
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Drug Discovery: Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.