Learn More
Eurofins Discoverx CLK4, active, 250 µg. (14-917M)
Supplier:  Eurofins Discoverx 14917M
Encompass
N-terminal 6xHis-tagged recombinant, human CLK4 amino acids 128-end. CLK4 protein belongs to the CDC2-like protein kinase (CLK) family. This protein kinase can interact with and phosphorylate the serine- and arginine-rich (SR) proteins, which are known to play an important role in the formation of spliceosomes, and thus may be involved in the regulation of alternative splicing. Studies in the Israeli sand Rat Psammomys obesus suggested that the ubiquitin-like 5 (UBL5/BEACON), a highly conserved ubiquitin-like protein, may interact with and regulate the activity of this kinase. Multiple alternatively spliced transcript variants have been observed, but the full length natures of which have not yet been determined. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9410