Learn More
Biotage CLMN 300UL VLM DD/BIOTIX 96
SDP
Supplier:  Biotage PTX930502
Encompass
Protein G PhyTip columns (300 µL volume) containing 5 µL affinity media per column, DD/Biotix 96               
Catalog No. NC3104416