Learn More
Aviva Systems Biology Corp CLMN Antibody - N-terminal region (ARP53773_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53773_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CLMN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CLMN. Alternate names: FLJ12383, KIAA1188ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61123