Learn More
Lakeland Industries CLEANMAX CLEAN NONSTERILE SMMS Cleanroom coverall with Hood. Bound seams, elastic back, wrists, ankles, thumbloops, zipper placket. Individually packaged. 25/case with overlay bag. Size 3X
Catalog No. NC3232450
Encompass_Preferred
CleanMax SELECT SMMS Cloth-like fabric Cleanroom Apparel is available in sterile and non-sterile configurations. Clean-Manufactured under strict process and environment controls in Lakeland owned, operated, and certified cleanrooms. CleanMax SELECT cleanroom garments meet IEST Particle Category II Cleanliness levels. CleanMax SELECT sterile products are validated to an SAL 10^-6 with sterile traceability down to the individual garment. 96% bacterial and particulate filtration makes CleanMax SELECT resistant to microbial, viral and blood borne contaminants, along with most API’s, and HD's. Easy donning fold, zipper plackets, elastic back in the coveralls, thumb loops and always bound seams provide a superior barrier to prevent breakthrough and strike through. Let Lakeland clean-manufactured CleanMax SELECT apparel be the cornerstone of your contamination control strategy. Coveralls are constructed with a fluid-repellant, low-linting, four layer breathable SMMS fabric.
Catalog No. NC3232450 Supplier Lakeland Industries Supplier No. SBC428CM3X
CleanMax SELECT SMMS Cloth-like fabric Cleanroom Apparel is available in sterile and non-sterile configurations. Clean-Manufactured under strict process and environment controls in Lakeland owned, operated, and certified cleanrooms. CleanMax SELECT cleanroom garments meet IEST Particle Category II Cleanliness levels. CleanMax SELECT sterile products are validated to an SAL 10^-6 with sterile traceability down to the individual garment. 96% bacterial and particulate filtration makes CleanMax SELECT resistant to microbial, viral and blood borne contaminants, along with most API’s, and HD's. Easy donning fold, zipper plackets, elastic back in the coveralls, thumb loops and always bound seams provide a superior barrier to prevent breakthrough and strike through. Let Lakeland clean-manufactured CleanMax SELECT apparel be the cornerstone of your contamination control strategy. Coveralls are constructed with a fluid-repellant, low-linting, four layer breathable SMMS fabric.