Learn More
Lakeland Industries CLEANMAX BULK PACK NonSterile SMMS Cleanroom coverall. Bound seams, elastic back, wrists, ankles, thumbloops, zipper placket. 25 per case with overlay bag. Size 4X
Catalog No. NC2945149
Encompass_Preferred
CleanMax SELECT SMMS Cleanroom BULK Apparel is available in non-sterile configurations only. Clean-Manufactured under strict process and environment controls in Lakeland owned, operated, and certified cleanrooms. CleanMax cleanroom garments meet IEST Cleanliness levels. 96% bacterial and particulate filtration makes CleanMax SELECT resistant to microbial, viral and blood borne contaminants, along w/ most API’s, HD’s, and some cytotoxic drugs. Easy donning fold, zipper plackets, elastic back in the coveralls, thumb loops and always bound seams provide a superior barrier to prevent breakthrough and strike through. Let Lakeland clean-manufactured CleanMax SELECT apparel be the cornerstone of your contamination control strategy. Coveralls are constructed w/ a fluid-repellant, low-linting, SMMS CLoth-like fabric. Bound neckline, zipper w/ placket, elastic wrist w/ thumb loops, elastic ankles and back for a more tailored fit. BULK PACKAGED IN OVERLAY BAG, 25/case.
Catalog No. NC2945149 Supplier Lakeland Industries Supplier No. SBC417CMB4X
CleanMax SELECT SMMS Cleanroom BULK Apparel is available in non-sterile configurations only. Clean-Manufactured under strict process and environment controls in Lakeland owned, operated, and certified cleanrooms. CleanMax cleanroom garments meet IEST Cleanliness levels. 96% bacterial and particulate filtration makes CleanMax SELECT resistant to microbial, viral and blood borne contaminants, along w/ most API’s, HD’s, and some cytotoxic drugs. Easy donning fold, zipper plackets, elastic back in the coveralls, thumb loops and always bound seams provide a superior barrier to prevent breakthrough and strike through. Let Lakeland clean-manufactured CleanMax SELECT apparel be the cornerstone of your contamination control strategy. Coveralls are constructed w/ a fluid-repellant, low-linting, SMMS CLoth-like fabric. Bound neckline, zipper w/ placket, elastic wrist w/ thumb loops, elastic ankles and back for a more tailored fit. BULK PACKAGED IN OVERLAY BAG, 25/case.