Learn More
Lakeland Industries CLEANMAX BULK PACK NonSterile SMMS Cleanroom coverall with Hood and Boot. Bound seams, elastic back, wrists, ankles, thumbloops, zipper placket. 25 per case with overlay bag. Size LG
Catalog No. NC3201730
Encompass_Preferred
available in non-sterile configurations only. Clean-Manufactured under strict process and environment controls in Lakeland owned, operated, and certified cleanrooms. CleanMax cleanroom garments meet IEST Cleanliness levels. 96% bacterial and particulate filtration makes CleanMax SELECT resistant to microbial, viral and blood borne contaminants, along with most API’s, HD’s, and some cytotoxic drugs. Easy donning fold, zipper plackets, elastic back in the coveralls, thumb loops and always bound seams provide a superior barrier to prevent breakthrough and strike through. Let Lakeland clean-manufactured CleanMax SELECT apparel be the cornerstone of your contamination control strategy. Coveralls are constructed with a fluid-repellant, low-linting, SMMS CLoth-like fabric. Built in hood and boots, zipper with placket, elastic wrist with thumb loops, elastic ankles and back for a more tailored fit. BULK PACKAGED IN OVERLAY BAG, 25/case.
Catalog No. NC3201730 Supplier Lakeland Industries Supplier No. SBC414CMBLG
available in non-sterile configurations only. Clean-Manufactured under strict process and environment controls in Lakeland owned, operated, and certified cleanrooms. CleanMax cleanroom garments meet IEST Cleanliness levels. 96% bacterial and particulate filtration makes CleanMax SELECT resistant to microbial, viral and blood borne contaminants, along with most API’s, HD’s, and some cytotoxic drugs. Easy donning fold, zipper plackets, elastic back in the coveralls, thumb loops and always bound seams provide a superior barrier to prevent breakthrough and strike through. Let Lakeland clean-manufactured CleanMax SELECT apparel be the cornerstone of your contamination control strategy. Coveralls are constructed with a fluid-repellant, low-linting, SMMS CLoth-like fabric. Built in hood and boots, zipper with placket, elastic wrist with thumb loops, elastic ankles and back for a more tailored fit. BULK PACKAGED IN OVERLAY BAG, 25/case.