Learn More
Aviva Systems Biology Corp CLN6 Antibody - middle region (ARP47371_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47371_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CLN6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CLN6. Alternate names: FLJ20561, HsT18960, nclf, CLN4AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-59526