Learn More
Lakeland Industries CLNMAX Non Strl Frock XL 30/CS
Catalog No. 502886369
Encompass_Preferred
CleanMax Microporous is available in sterile and non-sterile configurations. Clean-Manufactured in Lakeland owned and operated certified cleanrooms, under strict process and environmental controls CleanMax meets IEST Particle Category Cleanliness levels. CleanMax sterile products are validated to an SAL 10^-6 with sterile traceability down to the individual garment. Resistant to microbial, viral and blood borne contaminants, along with some cytotoxic drugs. Easy donning fold, zipper plackets, elastic back in the coveralls, thumb loops and always bound seams provide a superior barrier for the wearer. Frocks are constructed with microporous film laminate material that is fluid repellant and protects against breakthrough and strikethrough. Mandarin collar, zipper with placket, elastic wrist with thumb loops.
Catalog No. 50-288-6369 Supplier Lakeland Industries Supplier No. CTL191CMBXL
CleanMax Microporous is available in sterile and non-sterile configurations. Clean-Manufactured in Lakeland owned and operated certified cleanrooms, under strict process and environmental controls CleanMax meets IEST Particle Category Cleanliness levels. CleanMax sterile products are validated to an SAL 10^-6 with sterile traceability down to the individual garment. Resistant to microbial, viral and blood borne contaminants, along with some cytotoxic drugs. Easy donning fold, zipper plackets, elastic back in the coveralls, thumb loops and always bound seams provide a superior barrier for the wearer. Frocks are constructed with microporous film laminate material that is fluid repellant and protects against breakthrough and strikethrough. Mandarin collar, zipper with placket, elastic wrist with thumb loops.