Learn More
Lakeland Industries Lakeland CLEANMAX SELECT STERILE SMMS cleanroom frock. ISO Class 5-8 cleanrooms and all controlled environments. Clean manufactured. Individually packaged 30 per case with overlay bag. Size 3X
Catalog No. NC2887788
Encompass_Preferred
CleanMax Select SMMS is available in sterile and non-sterile configurations. Clean-Manufactured in Lakeland owned and operated certified cleanrooms, under strict process and environmental controls CleanMax Select meets IEST Particle Category Cleanliness levels. CleanMax Select SMMS sterile products are validated to an SAL 10^-6 with sterile traceability down to the individual garment. Resistant to microbial, viral and blood borne contaminants. Easy donning fold, zipper plackets, elastic back in the coveralls, thumb loops and always bound seams provide a superior barrier for the wearer. Designed to prevent breakthrough and strikethrough, let Lakeland clean-manufactured CleanMax Select be the cornerstone of contamination control strategy. Frocks are constructed with a four-layer SMMS cloth-like fabric. Mandarin collar, zipper with placket, elastic wrist with thumb loops. 30/case. Available in sizes small through 5 XL.
Catalog No. NC2887788 Supplier Lakeland Industries Supplier No. SBC191CS3X
CleanMax Select SMMS is available in sterile and non-sterile configurations. Clean-Manufactured in Lakeland owned and operated certified cleanrooms, under strict process and environmental controls CleanMax Select meets IEST Particle Category Cleanliness levels. CleanMax Select SMMS sterile products are validated to an SAL 10^-6 with sterile traceability down to the individual garment. Resistant to microbial, viral and blood borne contaminants. Easy donning fold, zipper plackets, elastic back in the coveralls, thumb loops and always bound seams provide a superior barrier for the wearer. Designed to prevent breakthrough and strikethrough, let Lakeland clean-manufactured CleanMax Select be the cornerstone of contamination control strategy. Frocks are constructed with a four-layer SMMS cloth-like fabric. Mandarin collar, zipper with placket, elastic wrist with thumb loops. 30/case. Available in sizes small through 5 XL.