Learn More
Lakeland Industries CLEANMAX CLEAN NONSTERILE Microporous Cleanroom coverall with Hood & Boot. Bound seams, elastic back, wrists, ankles, thumbloops, zipper placket. Individually packaged. 25/case with overlay bag. Size XL
Catalog No. NC3199685
Encompass_Preferred
Clean-Manufactured under strict process and environment controls in Lakeland owned, operated, and certified cleanrooms. CleanMax sterile products are validated to an SAL 10^-6 with sterile traceability down to the individual garment. 99.999% bacterial and particulate filtration makes CleanMax resistant to microbial, viral and blood borne contaminants, along with most API’s, HD’s, and some cytotoxic drugs. Easy donning fold, zipper plackets, elastic back in the coveralls, thumb loops and always bound seams provide a superior barrier to prevent breakthrough and strike through. Let Lakeland clean-manufactured CleanMax apparel be the cornerstone of your contamination control strategy. Coveralls are constructed with a fluid-repellant, low-linting, microporous fabric. Built in hood and boots, zipper with placket, elastic wrist with thumb loops, elastic ankles and back for a more tailored fit. Individually packaged, 25/case.
Catalog No. NC3199685 Supplier Lakeland Industries Supplier No. CTL414CMXL
Clean-Manufactured under strict process and environment controls in Lakeland owned, operated, and certified cleanrooms. CleanMax sterile products are validated to an SAL 10^-6 with sterile traceability down to the individual garment. 99.999% bacterial and particulate filtration makes CleanMax resistant to microbial, viral and blood borne contaminants, along with most API’s, HD’s, and some cytotoxic drugs. Easy donning fold, zipper plackets, elastic back in the coveralls, thumb loops and always bound seams provide a superior barrier to prevent breakthrough and strike through. Let Lakeland clean-manufactured CleanMax apparel be the cornerstone of your contamination control strategy. Coveralls are constructed with a fluid-repellant, low-linting, microporous fabric. Built in hood and boots, zipper with placket, elastic wrist with thumb loops, elastic ankles and back for a more tailored fit. Individually packaged, 25/case.