Learn More
Lakeland Industries CLEANMAX CLEAN NONSTERILE Microporous Cleanroom coverall. Bound seams, elastic back, wrists, ankles, thumbloops, zipper placket. Individually packaged. 25/case with overlay bag. Size 4X
Catalog No. NC2944969
Encompass_Preferred
Clean-Manufactured under strict process and environment controls in Lakeland owned, operated, and certified cleanrooms. CleanMax cleanroom garments meet IEST Particle Category I Cleanliness levels. CleanMax sterile products are validated to an SAL 10^-6 w/ sterile traceability down to the individual garment. 99.999% bacterial and particulate filtration makes CleanMax resistant to microbial, viral and blood borne contaminants, along w/ most API’s, HD’s, and some cytotoxic drugs. Easy donning fold, zipper plackets, elastic back in the coveralls, thumb loops and always bound seams provide a superior barrier to prevent breakthrough and strike through. Let Lakeland clean-manufactured CleanMax apparel be the cornerstone of your contamination control strategy. Coveralls are constructed w/ a fluid-repellant, low-linting, microporous fabric. Bound neckline, zipper w/ placket, elastic wrist w/ thumb loops, elastic ankles and back for a more tailored fit.
Catalog No. NC2944969 Supplier Lakeland Industries Supplier No. CTL417CM4X
Clean-Manufactured under strict process and environment controls in Lakeland owned, operated, and certified cleanrooms. CleanMax cleanroom garments meet IEST Particle Category I Cleanliness levels. CleanMax sterile products are validated to an SAL 10^-6 w/ sterile traceability down to the individual garment. 99.999% bacterial and particulate filtration makes CleanMax resistant to microbial, viral and blood borne contaminants, along w/ most API’s, HD’s, and some cytotoxic drugs. Easy donning fold, zipper plackets, elastic back in the coveralls, thumb loops and always bound seams provide a superior barrier to prevent breakthrough and strike through. Let Lakeland clean-manufactured CleanMax apparel be the cornerstone of your contamination control strategy. Coveralls are constructed w/ a fluid-repellant, low-linting, microporous fabric. Bound neckline, zipper w/ placket, elastic wrist w/ thumb loops, elastic ankles and back for a more tailored fit.