Learn More
Lakeland Industries CLEANMAX CLEAN NONSTERILE Microporous Cleanroom Hood with ties. Open face. Bound seams and oversized yoke. Individually packaged. 100/case with overlay bag. Size UNI
Catalog No. NC2957685
Encompass_Preferred
CleanMax Microporous Cleanroom Apparel is available in sterile and non-sterile configurations. Clean-Manufactured under strict process and environment controls in Lakeland owned, operated, and certified cleanrooms. CleanMax cleanroom garments meet IEST Particle Category I Cleanliness levels. CleanMax sterile products are validated to an SAL 10^-6 with sterile traceability down to the individual garment. 99.999% bacterial and particulate filtration makes CleanMax resistant to microbial, viral and blood borne contaminants, along with most API’s, HD’s, and some cytotoxic drugs. Easy donning fold, so wearers only touch the inside of the hood. Open face design to fall on chin to secure proper mask coverage. Bound seams provide a superior barrier to prevent breakthrough and strike through. Hoods are constructed with a PFA-free, fluid-repellant, low-linting, microporous fabric. Individually packaged, 100/case. Nonsterile. Unisize.
Catalog No. NC2957685 Supplier Lakeland Industries Supplier No. CTL713CM
CleanMax Microporous Cleanroom Apparel is available in sterile and non-sterile configurations. Clean-Manufactured under strict process and environment controls in Lakeland owned, operated, and certified cleanrooms. CleanMax cleanroom garments meet IEST Particle Category I Cleanliness levels. CleanMax sterile products are validated to an SAL 10^-6 with sterile traceability down to the individual garment. 99.999% bacterial and particulate filtration makes CleanMax resistant to microbial, viral and blood borne contaminants, along with most API’s, HD’s, and some cytotoxic drugs. Easy donning fold, so wearers only touch the inside of the hood. Open face design to fall on chin to secure proper mask coverage. Bound seams provide a superior barrier to prevent breakthrough and strike through. Hoods are constructed with a PFA-free, fluid-repellant, low-linting, microporous fabric. Individually packaged, 100/case. Nonsterile. Unisize.