Learn More
Lakeland Industries CLEANMAX CLEAN NONSTERILE Microporous Cleanroom Sleeves with thumbloops. Bound seams. Individually packaged in pairs. 50 pr/case with overlay bag. Size 18"
Catalog No. NC2957689
Encompass_Preferred
CleanMax Microporous Cleanroom Apparel is available in sterile and non-sterile configurations. Clean-Manufactured under strict process and environment controls in Lakeland owned, operated, and certified cleanrooms. CleanMax cleanroom garments meet IEST Particle Category I Cleanliness levels. CleanMax sterile products are validated to an SAL 10^-6 with sterile traceability down to the individual garment. 99.999% bacterial and particulate filtration makes CleanMax resistant to microbial, viral and blood borne contaminants, along with most API’s, HD’s, and some cytotoxic drugs. Easy donning fold so wearers only touch the inside of the sleeve. Thumb loops and bound seams provide a superior barrier to prevent breakthrough and strike through while securing the sleeve in place. Sleeves are constructed with a PFA-free, fluid-repellant, low-linting, microporous fabric. Encased Elastic on top and bottom. 18" in length. Individually packaged in pairs, 50 pr/case. Sterile
Catalog No. NC2957689 Supplier Lakeland Industries Supplier No. CTL850CMP18
CleanMax Microporous Cleanroom Apparel is available in sterile and non-sterile configurations. Clean-Manufactured under strict process and environment controls in Lakeland owned, operated, and certified cleanrooms. CleanMax cleanroom garments meet IEST Particle Category I Cleanliness levels. CleanMax sterile products are validated to an SAL 10^-6 with sterile traceability down to the individual garment. 99.999% bacterial and particulate filtration makes CleanMax resistant to microbial, viral and blood borne contaminants, along with most API’s, HD’s, and some cytotoxic drugs. Easy donning fold so wearers only touch the inside of the sleeve. Thumb loops and bound seams provide a superior barrier to prevent breakthrough and strike through while securing the sleeve in place. Sleeves are constructed with a PFA-free, fluid-repellant, low-linting, microporous fabric. Encased Elastic on top and bottom. 18" in length. Individually packaged in pairs, 50 pr/case. Sterile