Learn More
Lakeland Industries CLEANMAX BULK PACK NonSterile Microporous Cleanroom coverall with Hood. Bound seams, elastic back, wrists, ankles, thumbloops, zipper placket. 25 per case with overlay bag. Size MD
Catalog No. NC3232310
Encompass_Preferred
CleanMax Microporous Cleanroom BULK Apparel is available in non-sterile configurations only. Clean-Manufactured under strict process and environment controls in Lakeland owned, operated, and certified cleanrooms. CleanMax cleanroom garments meet IEST Cleanliness levels. 99.999% bacterial and particulate filtration makes CleanMax resistant to microbial, viral and blood borne contaminants, along with most API’s, HD’s, and some cytotoxic drugs. Easy donning fold, zipper plackets, elastic back in the coveralls, thumb loops and always bound seams provide a superior barrier to prevent breakthrough and strike through. Let Lakeland clean-manufactured CleanMax apparel be the cornerstone of your contamination control strategy. Coveralls are constructed with a fluid-repellant, low-linting, microporous fabric. Built in hood, zipper with placket, elastic wrist with thumb loops, elastic ankles and back for a more tailored fit. BULK PACKAGED IN OVERLAY BAG, 25/case.
Catalog No. NC3232310 Supplier Lakeland Industries Supplier No. CTL428CMBMD
CleanMax Microporous Cleanroom BULK Apparel is available in non-sterile configurations only. Clean-Manufactured under strict process and environment controls in Lakeland owned, operated, and certified cleanrooms. CleanMax cleanroom garments meet IEST Cleanliness levels. 99.999% bacterial and particulate filtration makes CleanMax resistant to microbial, viral and blood borne contaminants, along with most API’s, HD’s, and some cytotoxic drugs. Easy donning fold, zipper plackets, elastic back in the coveralls, thumb loops and always bound seams provide a superior barrier to prevent breakthrough and strike through. Let Lakeland clean-manufactured CleanMax apparel be the cornerstone of your contamination control strategy. Coveralls are constructed with a fluid-repellant, low-linting, microporous fabric. Built in hood, zipper with placket, elastic wrist with thumb loops, elastic ankles and back for a more tailored fit. BULK PACKAGED IN OVERLAY BAG, 25/case.