Learn More
Lakeland Industries CLEANMAX CLEAN NONSTERILE SMMS Cleanroom boot. Non-skid sole. 19" upper. Individually packaged in pairs. 50 pr/case with overlay bag. Size 2X
Supplier:  Lakeland Industries SBC903CMP2X
Encompass_Preferred
Clean-Manufactured under strict process and environment controls in Lakeland owned, operated, and certified cleanrooms. Let Lakeland clean-manufactured CleanMax SELECT apparel be the cornerstone of your contamination control strategy. CleanMax SELECT cleanroom garments meet IEST Particle Category II Cleanliness levels. CleanMax SELECT sterile products are validated to an SAL 10^-6 with sterile traceability down to the individual garment. 96% bacterial and particulate filtration makes CleanMax SELECT resistant to microbial, viral and blood borne contaminants, along with most API’s, and HD's. Easy donning fold, so wearers only touch the inside of the boot. Encased elastic, 19" upper and non-skid sole. Bound seams provide a superior barrier to prevent breakthrough and strike through. Boots are constructed with a PFA-free, fluid-repellant, low-linting, four layer SMMS fabric. Individually packaged in pairs, 50 pr/case. Nonsterile. Size 2X
Catalog No. NC2957733
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov