Learn More
Medchemexpress LLC Clobenpropit dihydrobromide | 145231-35-2 | MFCD00467655 | 99.8% | 470.65 g/mol | C14H19Br2ClN4S | 100 MG
SDP
Catalog No. 5000213192
Encompass_Preferred
Clobenpropit dihydrobromide is a research reagent used to study histamine receptor pharmacology and related signaling pathways. It is primarily a potent histamine H3 receptor antagonist/inverse agonist, with partial agonist activity at H4 receptors, and shows binding to serotonin 5-HT3 and α2A/α2C adrenoceptors. The compound is used in in vitro receptor profiling, apoptosis research, and preclinical studies of chemotherapeutic enhancement.
  • Potent H3 receptor antagonist/inverse agonist for receptor pharmacology studies.
  • Partial agonist activity at H4 receptors for selectivity experiments.
  • Binding to additional targets enables broader receptor profiling.
  • High purity (99.8%) for consistent experimental results.
  • Available as solid and solution formats for flexible dosing and formulation.
Catalog No. 50-002-13192 Supplier Medchemexpress LLC Supplier No. HY101198100MG
Clobenpropit dihydrobromide is a research reagent used to study histamine receptor pharmacology and related signaling pathways. It is primarily a potent histamine H3 receptor antagonist/inverse agonist, with partial agonist activity at H4 receptors, and shows binding to serotonin 5-HT3 and α2A/α2C adrenoceptors. The compound is used in in vitro receptor profiling, apoptosis research, and preclinical studies of chemotherapeutic enhancement.
  • Potent H3 receptor antagonist/inverse agonist for receptor pharmacology studies.
  • Partial agonist activity at H4 receptors for selectivity experiments.
  • Binding to additional targets enables broader receptor profiling.
  • High purity (99.8%) for consistent experimental results.
  • Available as solid and solution formats for flexible dosing and formulation.