Learn More
Medchemexpress LLC Clobenpropit dihydrobromide | 145231-35-2 | MFCD00467655 | 99.8% | 470.65 g/mol | C14H19Br2ClN4S | 25 MG
SDP
Catalog No. 5000211196
Encompass_Preferred
Clobenpropit dihydrobromide is a research-grade histamine H3 receptor antagonist and H4 receptor agonist supplied as the dihydrobromide salt. Provided as a high-purity solid, it is used in pharmacology and neuroscience research to probe histaminergic signaling, receptor pharmacology, and cellular responses in vitro. Consult the product datasheet and SDS for handling, storage, and safety information.
  • High-purity dihydrobromide salt suitable for research applications.
  • Relevant for histamine receptor pharmacology and signaling studies.
  • Stable solid form for accurate dosing in in vitro assays.
  • Compatible with standard solvent systems for reconstitution.
  • Offered in small pack sizes for laboratory-scale experiments.
Catalog No. 50-002-11196 Supplier Medchemexpress LLC Supplier No. HY10119825MG
Clobenpropit dihydrobromide is a research-grade histamine H3 receptor antagonist and H4 receptor agonist supplied as the dihydrobromide salt. Provided as a high-purity solid, it is used in pharmacology and neuroscience research to probe histaminergic signaling, receptor pharmacology, and cellular responses in vitro. Consult the product datasheet and SDS for handling, storage, and safety information.
  • High-purity dihydrobromide salt suitable for research applications.
  • Relevant for histamine receptor pharmacology and signaling studies.
  • Stable solid form for accurate dosing in in vitro assays.
  • Compatible with standard solvent systems for reconstitution.
  • Offered in small pack sizes for laboratory-scale experiments.