Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Clobenpropit dihydrobromide | 145231-35-2 | MFCD00467655 | 99.8% | 470.65 g/mol | C14H19Br2ClN4S | 50 MG
SDP

Catalog No. 5000211197
Encompass_Preferred

Clobenpropit dihydrobromide is a high-purity research chemical used as a potent histamine H3 receptor antagonist/inverse agonist and a partial H4 receptor agonist. Supplied as a white to off-white solid, it is intended for pharmacology and neuroscience studies to investigate histaminergic signaling and receptor pharmacology.

  • High reported purity (99.76%).
  • Molecular weight 470.65 g/mol; formula C14H19Br2ClN4S.
  • White to off-white solid form suitable for in vitro and in vivo research.
  • Recommended storage: 4°C sealed; in solvent -80°C up to 6 months.
  • Potent nanomolar activity at histamine H3 and partial activity at H4; reported off-target binding to 5-HT3 and α2 adrenoceptors.

Catalog No. 50-002-11197 Supplier Medchemexpress LLC Supplier No. HY10119850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Clobenpropit dihydrobromide is a high-purity research chemical used as a potent histamine H3 receptor antagonist/inverse agonist and a partial H4 receptor agonist. Supplied as a white to off-white solid, it is intended for pharmacology and neuroscience studies to investigate histaminergic signaling and receptor pharmacology.

  • High reported purity (99.76%).
  • Molecular weight 470.65 g/mol; formula C14H19Br2ClN4S.
  • White to off-white solid form suitable for in vitro and in vivo research.
  • Recommended storage: 4°C sealed; in solvent -80°C up to 6 months.
  • Potent nanomolar activity at histamine H3 and partial activity at H4; reported off-target binding to 5-HT3 and α2 adrenoceptors.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.