Learn More
Cayman Chemical Clobetasol PropIonate 10g
SDP
Catalog No. 502980991
Encompass_Preferred
A corticosteroid; binds to glucocorticoid receptors in a cell-free assay (IC50 = 3.17 nM) and inhibits proliferation of primary human skin fibroblasts at 5 UG/ml; topical administration reduces croton oil-induced EAr edema in mice
Catalog No. 50-298-0991 Supplier Cayman Chemical Supplier No. 2125110 G
A corticosteroid; binds to glucocorticoid receptors in a cell-free assay (IC50 = 3.17 nM) and inhibits proliferation of primary human skin fibroblasts at 5 UG/ml; topical administration reduces croton oil-induced EAr edema in mice