Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clobetasol propionate, ≥98%, MilliporeSigma™ Supelco™

Catalog No. 111012185 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2185 100 mg
11-101-2186 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-2185 Supplier MilliporeSigma Supelco Supplier No. C8037100MG
Add to Cart
Add to Cart

Clobetasol propionate is a novel and highly potent, topical corticosteroid, which is being assessed for its utility in the treatment of bullous pemphigoid.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Glass Bottle
CAS 25122-46-7
Physical Form Neat
Linear Formula C25H32 ClFO5
MDL Number MFCD00058499
Type Analytical Standard
Quantity 100 mg
Formula Weight 466.97
Percent Purity ≥98%
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.