Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Clobutinol (hydrochloride) | 1215-83-4 | 99.9% | 292.25 | 1 ML
SDP

Catalog No. 5000361588
Encompass_Preferred

Clobutinol hydrochloride is a compound with anti-tussive effects. It affects heart rate and blood pressure and can be used for cough-related research.

  • Anti-tussive effects
  • Affects heart rate and blood pressure
  • Used for cough-related research
  • 99.9% purity
  • Molecular weight: 292.25
  • CAS Number: 1215-83-4
  • Harmful if swallowed
  • Causes skin irritation
  • Causes serious eye irritation
  • May cause respiratory irritation

Catalog No. 50-003-61588 Supplier Medchemexpress LLC Supplier No. HY148144A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Clobutinol hydrochloride is a compound with anti-tussive effects. It affects heart rate and blood pressure and can be used for cough-related research.

  • Anti-tussive effects
  • Affects heart rate and blood pressure
  • Used for cough-related research
  • 99.9% purity
  • Molecular weight: 292.25
  • CAS Number: 1215-83-4
  • Harmful if swallowed
  • Causes skin irritation
  • Causes serious eye irritation
  • May cause respiratory irritation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.