Learn More
Cayman Chemical Clodesnitazene (citrate), Purity: ≥98%, 5 mg
SDP
Catalog No. 502945238
Encompass_Preferred
An analytical reference standard that is structurally similar to known opioids; intended for research and forensic applications
Catalog No. 50-294-5238 Supplier Cayman Chemical Supplier No. 426055 MG
An analytical reference standard that is structurally similar to known opioids; intended for research and forensic applications