Learn More
Medchemexpress LLC HY-136380 100mg Medchemexpress, Clodinafop-propargyl CAS:105512-06-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY136380100MG
Encompass_Preferred
Medchemexpress, HY-136380 100mg Clodinafop-propargyl CAS:105512-06-9 Clodinafop-propargyl, a main member of aryloxyphenoxy-propionate herbicides, is used for postemergence control of annual grasses in cereals, including Avena, Lolium, Setaria, Phalaris and Alopecurus spp. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1439