Learn More
Medchemexpress LLC Clodinafop-propargyl | 105512-06-9 | 98.0% | 349.74 | 10MM/1ML
SDP
Catalog No. 5000349132
Encompass_Preferred
Clodinafop-propargyl is a primary member of aryloxyphenoxy-propionate herbicides, utilized for the postemergence control of annual grasses in cereals. This compound also serves as a click chemistry reagent due to its alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • Used for postemergence control of annual grasses in cereals
  • Functions as a click chemistry reagent
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups
Catalog No. 50-003-49132 Supplier Medchemexpress LLC Supplier No. HY13638010MM/1ML
Clodinafop-propargyl is a primary member of aryloxyphenoxy-propionate herbicides, utilized for the postemergence control of annual grasses in cereals. This compound also serves as a click chemistry reagent due to its alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • Used for postemergence control of annual grasses in cereals
  • Functions as a click chemistry reagent
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups