Learn More
Medchemexpress LLC Clodronate disodium 250mg | 88416-50-6 | 250MG
SDP
Catalog No. 5000241321
Encompass_Preferred
Clodronate disodium tetrahydrate (Disodium clodronate tetrahydrate) is first-generation bisphosphonate with anti-osteoporotic anti-inflammatory and analgesic effects Clodronate disodium tetrahydrate is a selective potent reversible and Cl- competitive vesicular nucleotide transporter (VNUT) inhibitor with an IC50 of 15 6 nM Clodronate disodium tetrahydrate inhibits vesicular ATP release from neurons and reduces chronic neuropathic and inflammatory pain[1 [2
Catalog No. 50-002-41321 Supplier Medchemexpress LLC Supplier No. HY107794250MG
Clodronate disodium tetrahydrate (Disodium clodronate tetrahydrate) is first-generation bisphosphonate with anti-osteoporotic anti-inflammatory and analgesic effects Clodronate disodium tetrahydrate is a selective potent reversible and Cl- competitive vesicular nucleotide transporter (VNUT) inhibitor with an IC50 of 15 6 nM Clodronate disodium tetrahydrate inhibits vesicular ATP release from neurons and reduces chronic neuropathic and inflammatory pain[1 [2