Learn More
Selleck Chemical LLC Clodronate Disodium S4108-50mg
SDP
Catalog No. 503108614
Encompass_Preferred
Clodronate Disodium a bisphosphonate is a potent antiosteolytic agent which inhibits bone resorption
Catalog No. 50-310-8614 Supplier Selleck Chemical LLC Supplier No. S410850MG
Clodronate Disodium a bisphosphonate is a potent antiosteolytic agent which inhibits bone resorption