Learn More
Medchemexpress LLC Clodronate (disodium tetrahydrate) | 88416-50-6 | MFCD02940846 | 99.9% | 360.92 | CH10Cl2Na2O10P2 | 50 MG
SDP
Catalog No. 5000242336
Encompass_Preferred
Clodronate (disodium tetrahydrate) is a first-generation bisphosphonate research reagent used to study bone resorption, inflammatory and neuropathic pain mechanisms, and vesicular nucleotide transport. It is provided as a white to off-white solid with defined solubility and storage guidelines for laboratory use.
  • High purity (99.86%), suitable for research applications.
  • Solid powder form, white to off-white, easy to weigh and dissolve.
  • High water solubility (≈116.7 mg/mL) with protocols recommending ultrasonic warming.
  • Documented storage recommendations for sealed, moisture-free conditions.
  • Reported activity as a VNUT inhibitor that reduces vesicular ATP release.
Catalog No. 50-002-42336 Supplier Medchemexpress LLC Supplier No. HY10779450MG
Clodronate (disodium tetrahydrate) is a first-generation bisphosphonate research reagent used to study bone resorption, inflammatory and neuropathic pain mechanisms, and vesicular nucleotide transport. It is provided as a white to off-white solid with defined solubility and storage guidelines for laboratory use.
  • High purity (99.86%), suitable for research applications.
  • Solid powder form, white to off-white, easy to weigh and dissolve.
  • High water solubility (≈116.7 mg/mL) with protocols recommending ultrasonic warming.
  • Documented storage recommendations for sealed, moisture-free conditions.
  • Reported activity as a VNUT inhibitor that reduces vesicular ATP release.