Learn More
Medchemexpress LLC Clodronate (disodium tetrahydrate) | 88416-50-6 | MFCD02940846 | 99.9% | 360.92 g/mol | CH10Cl2Na2O10P2 | 1 ML
SDP
Catalog No. 5000240270
Encompass_Preferred
Clodronate (disodium tetrahydrate) is the disodium salt tetrahydrate of clodronic acid, a first-generation bisphosphonate used in biochemical and cell biology research to inhibit bone resorption, modulate macrophage populations, and study mineralization processes. It is supplied as solid material and as ready-to-use aqueous solutions for in vitro applications.
  • High reported purity (99.86%) suitable for research assays.
  • Available as a 10 mM aqueous solution and as solid quantities for flexibility.
  • Molecular weight 360.92 g/mol for precise dosing calculations.
  • Useful for studies of bone resorption, macrophage depletion, and calcification.
  • Includes SDS and COA documentation for quality control.
Catalog No. 50-002-40270 Supplier Medchemexpress LLC Supplier No. HY10779410MM 1ML
Clodronate (disodium tetrahydrate) is the disodium salt tetrahydrate of clodronic acid, a first-generation bisphosphonate used in biochemical and cell biology research to inhibit bone resorption, modulate macrophage populations, and study mineralization processes. It is supplied as solid material and as ready-to-use aqueous solutions for in vitro applications.
  • High reported purity (99.86%) suitable for research assays.
  • Available as a 10 mM aqueous solution and as solid quantities for flexibility.
  • Molecular weight 360.92 g/mol for precise dosing calculations.
  • Useful for studies of bone resorption, macrophage depletion, and calcification.
  • Includes SDS and COA documentation for quality control.