Learn More
Medchemexpress LLC Clodronate (disodium tetrahydrate) | 88416-50-6 | MFCD02940846 | 99.9% | 360.92 g/mol | CH10Cl2Na2O10P2 | 100 MG
SDP
Catalog No. 5000240263
Encompass_Preferred
Clodronate disodium tetrahydrate is a first-generation bisphosphonate research reagent used in studies of bone metabolism, inflammation, and analgesia. It is supplied as a white to off-white solid with high reported purity and accompanying analytical documents for laboratory use.
  • High purity (99.86%) suitable for analytical and biochemical applications.
  • White to off-white solid form for simple handling and weighing.
  • Provided with COA, SDS, and analytical data for quality assurance.
  • Available in small research pack sizes and as a ready-to-use solution.
  • Suitable as a reference standard in bone metabolism and bisphosphonate studies.
Catalog No. 50-002-40263 Supplier Medchemexpress LLC Supplier No. HY107794100MG
Clodronate disodium tetrahydrate is a first-generation bisphosphonate research reagent used in studies of bone metabolism, inflammation, and analgesia. It is supplied as a white to off-white solid with high reported purity and accompanying analytical documents for laboratory use.
  • High purity (99.86%) suitable for analytical and biochemical applications.
  • White to off-white solid form for simple handling and weighing.
  • Provided with COA, SDS, and analytical data for quality assurance.
  • Available in small research pack sizes and as a ready-to-use solution.
  • Suitable as a reference standard in bone metabolism and bisphosphonate studies.