Learn More
SAPPHIRE NORTH AMERICA CLODRONATE LIPOSOMES 5ML
Supplier: SAPPHIRE NORTH AMERICA C005
- Main component: Clodronate encapsulated within liposomes
- Volume: 5 mL
- Type: Multilamellar liposome suspension
- Suspension buffer: Phosphate Buffered Saline (PBS)
- Clodronate concentration: 5 mg/mL
- Max liposome size: 3 micron
- Average liposome size: 1.7 micron
- Lipid composition: Phosphatidylcholine and Cholesterol
- pH: 7.4
- Storage conditions: Store at 4°C, do not freeze
- Shelf life: 12 weeks after shipment
- Recommended use: For research use only
- Application: Macrophage depletion
- Administration methods: Intravenous, Intraperitoneal, Subcutaneous, Intranasal, Testicular injection
- Formulation condition: Prepared and packaged under sterile conditions
- Liposome charge: Slightly negatively charged
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.