Learn More
Cayman Chemical Clodronate (sodium salt hydrate), Purity: ≥95%, CAS: 88416-50-6, 50 mg
SDP
Catalog No. 502968756
Encompass_Preferred
A non-nitrogenous bisphosphonate that inhibits hydroxyapatite crystal growth (IC50 = 1.34 µM) and bone resorption; is cytotoxic to both osteoclasts and macrophages and has been used in biological research to selectively induce apoptosis of macrophage-like cells and cancer cells
Catalog No. 50-296-8756 Supplier Cayman Chemical Supplier No. 1613450 MG
A non-nitrogenous bisphosphonate that inhibits hydroxyapatite crystal growth (IC50 = 1.34 µM) and bone resorption; is cytotoxic to both osteoclasts and macrophages and has been used in biological research to selectively induce apoptosis of macrophage-like cells and cancer cells