Learn More
Selleck Chemical LLC clodronic acid
SDP
Catalog No. 5000003190
Encompass_Preferred
Clodronate disodium tetrahydrate (clodronic acid) is a diphosphonate which affects calcium metabolism It inhibits bone resorption and soft tissue calcification
Catalog No. 50-000-03190 Supplier Selleck Chemical LLC Supplier No. S566025MG
Add to Cart
Add to Cart
Clodronate disodium tetrahydrate (clodronic acid) is a diphosphonate which affects calcium metabolism It inhibits bone resorption and soft tissue calcification