Learn More
Medchemexpress LLC Clodronic acid disodium salt | 22560-50-5 | 100.5% | 100 MG
SDP
Catalog No. 5000407660
Encompass_Preferred
Clodronic acid disodium salt is a first-generation bisphosphonate that acts as an orally active osteoclastic bone resorption inhibitor. It is used in conditions characterized by high bone turnover, such as Paget's disease and osteolytic bone metastases.
  • Induces rapid apoptosis in osteoclasts
  • Prevents ADP translocation into mitochondria
  • Inhibits ATP production, leading to apoptosis
  • Demonstrated to slightly decrease hindpaw swelling in in vivo studies
Catalog No. 50-004-07660 Supplier Medchemexpress LLC Supplier No. HYB0657A100MG
Clodronic acid disodium salt is a first-generation bisphosphonate that acts as an orally active osteoclastic bone resorption inhibitor. It is used in conditions characterized by high bone turnover, such as Paget's disease and osteolytic bone metastases.
  • Induces rapid apoptosis in osteoclasts
  • Prevents ADP translocation into mitochondria
  • Inhibits ATP production, leading to apoptosis
  • Demonstrated to slightly decrease hindpaw swelling in in vivo studies