Learn More
TARGETMOL CHEMICALS INC Clodronic acid disodium salt
SDP
Catalog No. 502877100
Encompass_Preferred
Also available in 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Clodronic acid disodium salt (Lodronate), a bisphosphonate, is a potent antiosteolytic agent which inhibits bone resorption. Purity 100%
Catalog No. 50-287-7100 Supplier TARGETMOL CHEMICALS INC Supplier No. T6451500MG
Also available in 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Clodronic acid disodium salt (Lodronate), a bisphosphonate, is a potent antiosteolytic agent which inhibits bone resorption. Purity 100%