Learn More
Cayman Chemical Clofarabine, Purity: ≥98%, CAS: 123318-82-1, 50 mg
SDP
Catalog No. 502953576
Encompass_Preferred
A nucleoside analog that inhibits ribonucleotide reductase and DNA polymerase-α (IC50s = 65 and 3.9 nM, respectively) and is cytotoxic to K562 myelogenous leukemia cells (IC50 = 5 nM) as well as various other leukemias and solid tumors
Catalog No. 50-295-3576 Supplier Cayman Chemical Supplier No. 1412550 MG
A nucleoside analog that inhibits ribonucleotide reductase and DNA polymerase-α (IC50s = 65 and 3.9 nM, respectively) and is cytotoxic to K562 myelogenous leukemia cells (IC50 = 5 nM) as well as various other leukemias and solid tumors