Learn More
Apexbio Technology LLC Clofazimine 2030-63-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063387
Encompass_Preferred
Clofazimine is a rhimophenazine compound with demonstrated antimicrobial and anti-inflammatory properties commonly utilized in biomedical research related to tuberculosis treatment and drug-resistance studies It functions primarily through the intercalation into bacterial DNA interfering with microbial DNA replication and transcription processes Furthermore it may induce bacterial phospholipid membrane disruption and modulate inflammatory responses Experimental studies report IC50 values in the micromolar range against Mycobacterium tuberculosis strains Clofazimine is frequently employed as a reference compound in antimicrobial assays mechanistic studies on DNA-targeting agents and evaluations of cellular inflammatory pathways within infectious disease model systems
Catalog No. 50-000-63387 Supplier Apexbio Technology LLC Supplier No. B17105.1
Clofazimine is a rhimophenazine compound with demonstrated antimicrobial and anti-inflammatory properties commonly utilized in biomedical research related to tuberculosis treatment and drug-resistance studies It functions primarily through the intercalation into bacterial DNA interfering with microbial DNA replication and transcription processes Furthermore it may induce bacterial phospholipid membrane disruption and modulate inflammatory responses Experimental studies report IC50 values in the micromolar range against Mycobacterium tuberculosis strains Clofazimine is frequently employed as a reference compound in antimicrobial assays mechanistic studies on DNA-targeting agents and evaluations of cellular inflammatory pathways within infectious disease model systems